Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCGGCCGAGCGCTTTCTTTCGT[C/T]GTTTTCCAAGGGGGTTTTAAAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610129 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHST12 - carbohydrate (chondroitin 4) sulfotransferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243794.1 | Intron | NP_001230723.1 | ||||
NM_001243795.1 | Intron | NP_001230724.1 | ||||
NM_018641.4 | Intron | NP_061111.1 | ||||
XM_011515443.2 | Intron | XP_011513745.1 | ||||
XM_011515444.2 | Intron | XP_011513746.1 |
Set Membership: |
HapMap |