Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602504 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SHOX2 - short stature homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163678.1 | Intron | NP_001157150.1 | ||||
NM_003030.4 | Intron | NP_003021.3 | ||||
NM_006884.3 | Intron | NP_006875.2 | ||||
XM_006713727.3 | Intron | XP_006713790.1 | ||||
XM_006713728.3 | Intron | XP_006713791.1 | ||||
XM_017007053.1 | Intron | XP_016862542.1 | ||||
XM_017007054.1 | Intron | XP_016862543.1 | ||||
XM_017007055.1 | Intron | XP_016862544.1 |
Set Membership: |
Validated |