Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTGGGCTGCATGAGGAGCCTCC[C/T]ACATCCCAGAAGAGGTGGGGGTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138079 MIM: 613993 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCK PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCK - glucokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL7 - myosin light chain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021223.2 | Intron | NP_067046.1 | ||||
XM_005249817.4 | Intron | XP_005249874.1 | ||||
XM_011515463.2 | Intron | XP_011513765.1 | ||||
XM_011515464.2 | Intron | XP_011513766.1 | ||||
XM_011515465.2 | Intron | XP_011513767.1 | ||||
XM_017012478.1 | Intron | XP_016867967.1 | ||||
XM_017012479.1 | Intron | XP_016867968.1 |
Set Membership: |
HapMap |