Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAGAAACACGCATGAACTTTTC[C/T]ATAACCTTGATATATGGAAAGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612136 MIM: 612137 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ECHDC1 - ethylmalonyl-CoA decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002030.1 | Intron | NP_001002030.1 | ||||
NM_001105544.1 | Intron | NP_001099014.1 | ||||
NM_001105545.1 | Intron | NP_001099015.1 | ||||
NM_001139510.1 | Intron | NP_001132982.1 | ||||
NM_018479.3 | Intron | NP_060949.2 | ||||
XM_005267047.2 | Intron | XP_005267104.1 | ||||
XM_005267048.1 | Intron | XP_005267105.1 | ||||
XM_005267049.2 | Intron | XP_005267106.1 | ||||
XM_005267050.2 | Intron | XP_005267107.1 | ||||
XM_011535942.2 | Intron | XP_011534244.1 | ||||
XM_011535943.2 | Intron | XP_011534245.1 | ||||
XM_017011056.1 | Intron | XP_016866545.1 |
RNF146 - ring finger protein 146 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |