Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAAATGGCAGGTGCTGTTTGCTA[C/T]TGAAAGTACTGGAAGGCTGGGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 138600 MIM: 138610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317950.1 | Intron | NP_001304879.1 | ||||
NM_001317952.1 | Intron | NP_001304881.1 | ||||
NM_030767.5 | Intron | NP_110394.3 | ||||
XM_005252244.2 | Intron | XP_005252301.1 | ||||
XM_005252245.1 | Intron | XP_005252302.1 | ||||
XM_005252247.4 | Intron | XP_005252304.1 | ||||
XM_005252248.2 | Intron | XP_005252305.1 | ||||
XM_006717294.1 | Intron | XP_006717357.1 | ||||
XM_006717295.2 | Intron | XP_006717358.1 | ||||
XM_011519063.2 | Intron | XP_011517365.1 | ||||
XM_011519064.2 | Intron | XP_011517366.1 | ||||
XM_011519065.2 | Intron | XP_011517367.2 | ||||
XM_011519066.2 | Intron | XP_011517368.1 | ||||
XM_017015172.1 | Intron | XP_016870661.1 |
ORM1 - orosomucoid 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORM2 - orosomucoid 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |