Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609045 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR141 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR141 - G protein-coupled receptor 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011515374.2 | 125 | Intron | XP_011513676.1 | |||
XM_011515375.2 | 125 | Intron | XP_011513677.1 | |||
XM_011515383.2 | 125 | Intron | XP_011513685.1 | |||
XM_011515385.2 | 125 | Intron | XP_011513687.1 | |||
XM_011515386.2 | 125 | Intron | XP_011513688.1 | |||
XM_011515388.2 | 125 | UTR 5 | XP_011513690.1 | |||
XM_011515389.2 | 125 | Intron | XP_011513691.1 | |||
XM_017012160.1 | 125 | Intron | XP_016867649.1 | |||
XM_017012161.1 | 125 | Intron | XP_016867650.1 | |||
XM_017012162.1 | 125 | Intron | XP_016867651.1 | |||
XM_017012163.1 | 125 | Intron | XP_016867652.1 | |||
XM_017012164.1 | 125 | Intron | XP_016867653.1 | |||
XM_017012165.1 | 125 | Intron | XP_016867654.1 | |||
XM_017012166.1 | 125 | Intron | XP_016867655.1 | |||
XM_017012167.1 | 125 | Intron | XP_016867656.1 |
Set Membership: |
HapMap |