Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCACAGGATGCCCGACACACAC[A/G]CTTCTCTAGTTTTGGTGTAGACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605712 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPTLC1 - serine palmitoyltransferase long chain base subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281303.1 | Intron | NP_001268232.1 | ||||
NM_006415.3 | Intron | NP_006406.1 | ||||
NM_178324.2 | Intron | NP_847894.1 | ||||
XM_011518138.2 | Intron | XP_011516440.1 | ||||
XM_011518139.2 | Intron | XP_011516441.1 | ||||
XM_017014200.1 | Intron | XP_016869689.1 | ||||
XM_017014201.1 | Intron | XP_016869690.1 | ||||
XM_017014202.1 | Intron | XP_016869691.1 |
Set Membership: |
HapMap |