Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCTGACAGCAAAAACATACTC[C/T]TTGAGGGAGGGGAGCCCATTGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605513 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA2026 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KIAA2026 - KIAA2026 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017969.2 | Intron | NP_001017969.2 | ||||
XM_005251385.2 | Intron | XP_005251442.1 | ||||
XM_006716730.3 | Intron | XP_006716793.1 | ||||
XM_011517759.2 | Intron | XP_011516061.1 | ||||
XM_011517760.2 | Intron | XP_011516062.1 | ||||
XM_011517761.2 | Intron | XP_011516063.1 | ||||
XM_017014331.1 | Intron | XP_016869820.1 | ||||
XM_017014332.1 | Intron | XP_016869821.1 | ||||
XM_017014333.1 | Intron | XP_016869822.1 | ||||
XM_017014334.1 | Intron | XP_016869823.1 | ||||
XM_017014335.1 | Intron | XP_016869824.1 | ||||
XM_017014336.1 | Intron | XP_016869825.1 |
MLANA - melan-A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005511.1 | Intron | NP_005502.1 |