Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCATATTATACCCTTATTTAGC[G/T]CCCAGCTCCATCAGCTCATATTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608040 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT74 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IFT74 - intraflagellar transport 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099222.1 | 2988 | Intron | NP_001092692.1 | |||
NM_001099223.1 | 2988 | Intron | NP_001092693.1 | |||
NM_001099224.1 | 2988 | Intron | NP_001092694.1 | |||
NM_025103.2 | 2988 | Intron | NP_079379.2 | |||
XM_011518035.2 | 2988 | Intron | XP_011516337.1 | |||
XM_011518036.2 | 2988 | Intron | XP_011516338.1 | |||
XM_017015163.1 | 2988 | Intron | XP_016870652.1 | |||
XM_017015164.1 | 2988 | Intron | XP_016870653.1 |
LRRC19 - leucine rich repeat containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022901.2 | 2988 | UTR 3 | NP_075052.1 |