Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCAGTTTAAATGCTGATAATTA[C/T]TGTGAAATGGCACACTAAAGAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613492 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACER2 - alkaline ceramidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010887.2 | Intron | NP_001010887.2 | ||||
XM_005251448.3 | Intron | XP_005251505.1 | ||||
XM_011517858.1 | Intron | XP_011516160.1 | ||||
XM_011517859.2 | Intron | XP_011516161.1 | ||||
XM_017014694.1 | Intron | XP_016870183.1 |