Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612805 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6878 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6878 - microRNA 6878 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARS2 - threonyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271895.1 | Intron | NP_001258824.1 | ||||
NM_001271896.1 | Intron | NP_001258825.1 | ||||
NM_025150.4 | Intron | NP_079426.2 | ||||
XM_006711555.1 | Intron | XP_006711618.1 | ||||
XM_006711556.1 | Intron | XP_006711619.1 | ||||
XM_017002394.1 | Intron | XP_016857883.1 | ||||
XM_017002395.1 | Intron | XP_016857884.1 |
Set Membership: |
HapMap |