Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCAGAGTCTTACATCCCAATAA[A/G]TTAAAAGCCAAAGTGAATATAATTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610035 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VPS45 - vacuolar protein sorting 45 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279353.1 | Intron | NP_001266282.1 | ||||
NM_001279354.1 | Intron | NP_001266283.1 | ||||
NM_001279355.1 | Intron | NP_001266284.1 | ||||
NM_007259.4 | Intron | NP_009190.2 | ||||
XM_017000164.1 | Intron | XP_016855653.1 |
Set Membership: |
HapMap |