Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCCTAGGCCTCCACAGGAGCA[A/G]GTGGGGCCTCTGATGGTAAAAGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194510 MIM: 601261 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF3 - zinc finger protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN21 - zinc finger and SCAN domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145914.2 | 204 | Silent Mutation | CAA,CAG | Q,Q 20 | NP_666019.1 | |
XM_005250568.4 | 204 | Silent Mutation | CAA,CAG | Q,Q 20 | XP_005250625.1 | |
XM_006716113.3 | 204 | Silent Mutation | CAA,CAG | Q,Q 20 | XP_006716176.1 | |
XM_017012584.1 | 204 | Silent Mutation | CAA,CAG | Q,Q 20 | XP_016868073.1 | |
XM_017012585.1 | 204 | Silent Mutation | CAA,CAG | Q,Q 20 | XP_016868074.1 |
Set Membership: |
HapMap Validated |