Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604684 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42SE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC42SE1 - CDC42 small effector 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GABPB2 - GA binding protein transcription factor beta subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLLT11 - myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006818.3 | Intron | NP_006809.1 |