Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTTTGTTGCTTGTTTAGAAAT[A/G]TATCCCTTGTTTTACCTCAGATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604538 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF2C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIF2C - kinesin family member 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297655.1 | Intron | NP_001284584.1 | ||||
NM_001297656.1 | Intron | NP_001284585.1 | ||||
NM_001297657.1 | Intron | NP_001284586.1 | ||||
NM_006845.3 | Intron | NP_006836.2 | ||||
XM_011540540.2 | Intron | XP_011538842.1 | ||||
XM_011540541.2 | Intron | XP_011538843.1 |
Set Membership: |
HapMap |