Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRCP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
||||||
SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CRCP - CGRP receptor component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040647.1 | Intron | NP_001035737.1 | ||||
NM_001040648.1 | Intron | NP_001035738.1 | ||||
NM_001142414.1 | Intron | NP_001135886.1 | ||||
NM_014478.4 | Intron | NP_055293.1 |
Set Membership: |
HapMap Validated |