Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM32 - ADAM metallopeptidase domain 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313994.1 | Intron | NP_001300923.1 | ||||
NM_145004.6 | Intron | NP_659441.3 | ||||
XM_006716298.2 | Intron | XP_006716361.1 | ||||
XM_006716299.1 | Intron | XP_006716362.1 | ||||
XM_011544433.1 | Intron | XP_011542735.1 | ||||
XM_011544434.1 | Intron | XP_011542736.1 | ||||
XM_017013183.1 | Intron | XP_016868672.1 | ||||
XM_017013184.1 | Intron | XP_016868673.1 | ||||
XM_017013185.1 | Intron | XP_016868674.1 | ||||
XM_017013186.1 | Intron | XP_016868675.1 | ||||
XM_017013187.1 | Intron | XP_016868676.1 | ||||
XM_017013188.1 | Intron | XP_016868677.1 |
Set Membership: |
HapMap |