Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAATTCAGTCTCAGAGTTTCA[C/T]GCAAGCTCTCTCCTATTGCACTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THUMPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THUMPD1 - THUMP domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304550.1 | Intron | NP_001291479.1 | ||||
NM_017736.4 | Intron | NP_060206.2 | ||||
XM_011545891.1 | Intron | XP_011544193.1 | ||||
XM_017023433.1 | Intron | XP_016878922.1 |