Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTACATCACCCAGAGACCCCTCA[G/A]TAACTAACCAGACTGTATCCCCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604756 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF273 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF273 - zinc finger protein 273 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |