Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTAGGTGAACGTAACAGCTTAC[C/T]ATTTTTGTTTGCTGAGCTCTCCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609453 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GOLGA7 - golgin A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002296.1 | Intron | NP_001002296.1 | ||||
NM_001174124.1 | Intron | NP_001167595.1 | ||||
NM_016099.2 | Intron | NP_057183.2 |