Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTAGCTCTCTCTCCTCCTGGG[C/T]CTCAGGGCAACCCCTGCTTTAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614397 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MFSD2A - major facilitator superfamily domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136493.2 | Intron | NP_001129965.1 | ||||
NM_001287808.1 | Intron | NP_001274737.1 | ||||
NM_001287809.1 | Intron | NP_001274738.1 | ||||
NM_032793.4 | Intron | NP_116182.2 | ||||
XM_005271285.1 | Intron | XP_005271342.1 | ||||
XM_011542312.2 | Intron | XP_011540614.1 |