Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615730 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK7 - dedicator of cytokinesis 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271999.1 | Intron | NP_001258928.1 | ||||
NM_001272000.1 | Intron | NP_001258929.1 | ||||
NM_001272001.1 | Intron | NP_001258930.1 | ||||
NM_001272002.1 | Intron | NP_001258931.1 | ||||
NM_033407.3 | Intron | NP_212132.2 | ||||
XM_005271292.2 | Intron | XP_005271349.1 | ||||
XM_011542326.2 | Intron | XP_011540628.1 | ||||
XM_011542327.2 | Intron | XP_011540629.1 | ||||
XM_011542328.2 | Intron | XP_011540630.1 | ||||
XM_011542330.2 | Intron | XP_011540632.1 | ||||
XM_017002639.1 | Intron | XP_016858128.1 | ||||
XM_017002640.1 | Intron | XP_016858129.1 |
Set Membership: |
HapMap |