Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601212 MIM: 617007 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTK2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTK2B - protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004103.4 | Intron | NP_004094.3 | ||||
NM_173174.2 | Intron | NP_775266.1 | ||||
NM_173175.2 | Intron | NP_775267.1 | ||||
NM_173176.2 | Intron | NP_775268.1 | ||||
XM_005273447.4 | Intron | XP_005273504.1 | ||||
XM_005273448.4 | Intron | XP_005273505.1 | ||||
XM_011544441.2 | Intron | XP_011542743.1 | ||||
XM_011544442.2 | Intron | XP_011542744.1 | ||||
XM_017013214.1 | Intron | XP_016868703.1 | ||||
XM_017013215.1 | Intron | XP_016868704.1 | ||||
XM_017013216.1 | Intron | XP_016868705.1 |
TRIM35 - tripartite motif containing 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |