Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGATTGAGAAGGTGGAATTTGAT[C/T]GGAATAACTGGATCATCTCACGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603302 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY9 - adenylate cyclase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001116.3 | Intron | NP_001107.2 | ||||
XM_005255079.3 | Intron | XP_005255136.1 | ||||
XM_011522353.2 | Intron | XP_011520655.1 |
LOC102724927 - uncharacterized LOC102724927 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |