Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300654 MIM: 300621 MIM: 300517 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAAH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAAH2 - fatty acid amide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174912.3 | Intron | NP_777572.2 | ||||
XM_005261982.1 | Intron | XP_005262039.1 | ||||
XM_005261985.2 | Intron | XP_005262042.1 | ||||
XM_005261986.3 | Intron | XP_005262043.1 | ||||
XM_005261987.3 | Intron | XP_005262044.1 | ||||
XM_011530767.2 | Intron | XP_011529069.1 | ||||
XM_017029290.1 | Intron | XP_016884779.1 | ||||
XM_017029291.1 | Intron | XP_016884780.1 | ||||
XM_017029292.1 | Intron | XP_016884781.1 | ||||
XM_017029293.1 | Intron | XP_016884782.1 | ||||
XM_017029294.1 | Intron | XP_016884783.1 |
SPIN2A - spindlin family member 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIN2B - spindlin family member 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |