Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAGATCCCCAAGACGATCTTAC[A/C]CTTCAGGGATTCCCACACTATCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605448 MIM: 610820 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RUNDC3A - RUN domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144825.1 | Intron | NP_001138297.1 | ||||
NM_001144826.1 | Intron | NP_001138298.1 | ||||
NM_006695.4 | Intron | NP_006686.1 | ||||
XM_005256964.3 | Intron | XP_005257021.1 | ||||
XM_017024035.1 | Intron | XP_016879524.1 | ||||
XM_017024036.1 | Intron | XP_016879525.1 | ||||
XM_017024037.1 | Intron | XP_016879526.1 | ||||
XM_017024038.1 | Intron | XP_016879527.1 | ||||
XM_017024039.1 | Intron | XP_016879528.1 | ||||
XM_017024040.1 | Intron | XP_016879529.1 |
RUNDC3A-AS1 - RUNDC3A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A39 - solute carrier family 25 member 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |