Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610673 MIM: 608745 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NAIF1 - nuclear apoptosis inducing factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A25 - solute carrier family 25 member 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006641.3 | Intron | NP_001006642.1 | ||||
NM_001006642.3 | Intron | NP_001006643.1 | ||||
NM_001265614.2 | Intron | NP_001252543.1 | ||||
NM_052901.4 | Intron | NP_443133.2 | ||||
XM_005251688.1 | Intron | XP_005251745.1 | ||||
XM_005251689.4 | Intron | XP_005251746.1 | ||||
XM_006716948.2 | Intron | XP_006717011.1 | ||||
XM_006716949.3 | Intron | XP_006717012.1 |
Set Membership: |
HapMap Validated |