Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605882 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BRIP1 - BRCA1 interacting protein C-terminal helicase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032043.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1137 | NP_114432.2 | |
XM_011525332.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1157 | XP_011523634.1 | |
XM_011525333.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1157 | XP_011523635.1 | |
XM_011525334.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1157 | XP_011523636.1 | |
XM_011525335.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1137 | XP_011523637.1 | |
XM_011525336.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1117 | XP_011523638.1 | |
XM_011525337.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 1090 | XP_011523639.1 | |
XM_011525338.2 | 4208 | Silent Mutation | TAC,TAT | Y,Y 996 | XP_011523640.1 | |
XM_011525339.2 | 4208 | Intron | XP_011523641.1 | |||
XM_011525340.2 | 4208 | Intron | XP_011523642.1 | |||
XM_011525341.2 | 4208 | Intron | XP_011523643.1 | |||
XM_017025200.1 | 4208 | Silent Mutation | TAC,TAT | Y,Y 976 | XP_016880689.1 | |
XM_017025201.1 | 4208 | Silent Mutation | TAC,TAT | Y,Y 976 | XP_016880690.1 | |
XM_017025202.1 | 4208 | Silent Mutation | TAC,TAT | Y,Y 519 | XP_016880691.1 | |
XM_017025203.1 | 4208 | Silent Mutation | TAC,TAT | Y,Y 519 | XP_016880692.1 |
Set Membership: |
HapMap Validated |