Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGGAGGGCGGGGTGGGGTTTGCT[A/G]TAAGGGTGGGTCAAGGCCAGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615526 MIM: 163260 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPZ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COPZ2 - coatomer protein complex subunit zeta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016429.3 | Intron | NP_057513.1 | ||||
XM_011524874.2 | Intron | XP_011523176.1 |
NFE2L1 - nuclear factor, erythroid 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003204.2 | Intron | NP_003195.1 | ||||
XM_005257410.3 | Intron | XP_005257467.1 | ||||
XM_005257411.3 | Intron | XP_005257468.1 | ||||
XM_005257412.3 | Intron | XP_005257469.1 | ||||
XM_005257413.3 | Intron | XP_005257470.1 | ||||
XM_005257415.3 | Intron | XP_005257472.1 | ||||
XM_017024691.1 | Intron | XP_016880180.1 |
Set Membership: |
HapMap |