Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGATGACTTGTTTTTTCCTTG[C/T]ATCTTGCCTTTTAAACATTATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613240 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1R1C - protein phosphatase 1 regulatory inhibitor subunit 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080545.2 | Intron | NP_001074014.1 | ||||
NM_001261424.1 | Intron | NP_001248353.1 | ||||
NM_001261425.1 | Intron | NP_001248354.1 | ||||
XM_017003451.1 | Intron | XP_016858940.1 |
Set Membership: |
HapMap |