Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603086 MIM: 607607 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ART3 - ADP-ribosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP54 - nucleoporin 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278603.1 | 2033 | UTR 3 | NP_001265532.1 | |||
NM_017426.3 | 2033 | UTR 3 | NP_059122.2 | |||
XM_006714237.1 | 2033 | UTR 3 | XP_006714300.1 | |||
XM_011532033.2 | 2033 | UTR 3 | XP_011530335.1 | |||
XM_011532034.2 | 2033 | UTR 3 | XP_011530336.1 | |||
XM_017008294.1 | 2033 | UTR 3 | XP_016863783.1 | |||
XM_017008295.1 | 2033 | UTR 3 | XP_016863784.1 |
Set Membership: |
HapMap |