Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
G3BP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
G3BP2 - G3BP stress granule assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012297.4 | 2608 | UTR 3 | NP_036429.2 | |||
NM_203504.2 | 2608 | UTR 3 | NP_987100.1 | |||
NM_203505.2 | 2608 | UTR 3 | NP_987101.1 | |||
XM_005263382.2 | 2608 | UTR 3 | XP_005263439.1 | |||
XM_005263383.3 | 2608 | UTR 3 | XP_005263440.1 | |||
XM_011532441.2 | 2608 | UTR 3 | XP_011530743.1 | |||
XM_017008876.1 | 2608 | UTR 3 | XP_016864365.1 | |||
XM_017008877.1 | 2608 | UTR 3 | XP_016864366.1 | |||
XM_017008878.1 | 2608 | UTR 3 | XP_016864367.1 | |||
XM_017008879.1 | 2608 | UTR 3 | XP_016864368.1 |