Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTTTAGAGTGAATATTTCTTA[A/G]GTCTTTAGTATGCATATGAGGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607550 | ||||||||||||||||||||
Literature Links: |
SLC16A10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC16A10 - solute carrier family 16 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018593.4 | Intron | NP_061063.2 | ||||
XM_006715329.2 | Intron | XP_006715392.1 | ||||
XM_011535422.2 | Intron | XP_011533724.1 | ||||
XM_017010237.1 | Intron | XP_016865726.1 |