Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF385D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF385D - zinc finger protein 385D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024697.2 | 2681 | Intron | NP_078973.1 | |||
XM_011534122.1 | 2681 | Intron | XP_011532424.1 | |||
XM_011534123.2 | 2681 | Intron | XP_011532425.1 | |||
XM_011534124.2 | 2681 | Intron | XP_011532426.1 | |||
XM_017007191.1 | 2681 | UTR 3 | XP_016862680.1 | |||
XM_017007192.1 | 2681 | UTR 3 | XP_016862681.1 | |||
XM_017007193.1 | 2681 | UTR 3 | XP_016862682.1 | |||
XM_017007194.1 | 2681 | UTR 3 | XP_016862683.1 | |||
XM_017007195.1 | 2681 | UTR 3 | XP_016862684.1 | |||
XM_017007196.1 | 2681 | UTR 3 | XP_016862685.1 | |||
XM_017007197.1 | 2681 | UTR 3 | XP_016862686.1 | |||
XM_017007198.1 | 2681 | UTR 3 | XP_016862687.1 | |||
XM_017007199.1 | 2681 | UTR 3 | XP_016862688.1 | |||
XM_017007200.1 | 2681 | UTR 3 | XP_016862689.1 | |||
XM_017007201.1 | 2681 | UTR 3 | XP_016862690.1 | |||
XM_017007202.1 | 2681 | UTR 3 | XP_016862691.1 | |||
XM_017007203.1 | 2681 | Intron | XP_016862692.1 |