Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HGD - homogentisate 1,2-dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABL3 - RAB, member of RAS oncogene family like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173825.3 | 1233 | UTR 3 | NP_776186.2 | |||
XM_006713595.2 | 1233 | UTR 3 | XP_006713658.1 | |||
XM_017006227.1 | 1233 | UTR 3 | XP_016861716.1 | |||
XM_017006228.1 | 1233 | UTR 3 | XP_016861717.1 |