Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606045 MIM: 603574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB12 - EF-hand calcium binding domain 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFT122 - intraflagellar transport 122 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MBD4 - methyl-CpG binding domain 4, DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276270.1 | 1954 | Silent Mutation | GGA,GGG | G,G 530 | NP_001263199.1 | |
NM_001276271.1 | 1954 | Silent Mutation | GGA,GGG | G,G 536 | NP_001263200.1 | |
NM_001276272.1 | 1954 | Silent Mutation | GGA,GGG | G,G 536 | NP_001263201.1 | |
NM_001276273.1 | 1954 | Silent Mutation | GGA,GGG | G,G 218 | NP_001263202.1 | |
NM_003925.2 | 1954 | Silent Mutation | GGA,GGG | G,G 536 | NP_003916.1 | |
XM_011513268.2 | 1954 | Silent Mutation | GGA,GGG | G,G 141 | XP_011511570.1 |