Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606993 MIM: 614461 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IP6K3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IP6K3 - inositol hexakisphosphate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142883.1 | 2637 | UTR 3 | NP_001136355.1 | |||
NM_054111.4 | 2637 | UTR 3 | NP_473452.2 | |||
XM_005248842.3 | 2637 | UTR 3 | XP_005248899.1 | |||
XM_005248843.3 | 2637 | UTR 3 | XP_005248900.1 | |||
XM_011514295.2 | 2637 | Intron | XP_011512597.1 |
UQCC2 - ubiquinol-cytochrome c reductase complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |