Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300051 MIM: 300170 | ||||||||||||||||||||
Literature Links: |
GPM6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPM6B - glycoprotein M6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001994.2 | 2262 | UTR 3 | NP_001001994.1 | |||
NM_001001995.2 | 2262 | Intron | NP_001001995.1 | |||
NM_001001996.2 | 2262 | UTR 3 | NP_001001996.1 | |||
NM_001318729.1 | 2262 | Intron | NP_001305658.1 | |||
NM_005278.4 | 2262 | UTR 3 | NP_005269.1 | |||
XM_005274489.4 | 2262 | Intron | XP_005274546.1 | |||
XM_011545497.2 | 2262 | Intron | XP_011543799.1 | |||
XM_017029432.1 | 2262 | UTR 3 | XP_016884921.1 | |||
XM_017029433.1 | 2262 | Intron | XP_016884922.1 | |||
XM_017029434.1 | 2262 | UTR 3 | XP_016884923.1 |
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003611.2 | 2262 | Intron | NP_003602.1 |