Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2CD5 - C2 calcium dependent domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286173.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1020 | NP_001273102.1 | |
NM_001286174.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1018 | NP_001273103.1 | |
NM_001286175.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1021 | NP_001273104.1 | |
NM_001286176.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1018 | NP_001273105.1 | |
NM_001286177.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1009 | NP_001273106.1 | |
NM_014802.2 | 2325 | Silent Mutation | GAC,GAT | D,D 967 | NP_055617.1 | |
XM_005253538.2 | 2325 | Silent Mutation | GAC,GAT | D,D 958 | XP_005253595.1 | |
XM_005253539.3 | 2325 | Silent Mutation | GAC,GAT | D,D 640 | XP_005253596.1 | |
XM_006719170.2 | 2325 | Silent Mutation | GAC,GAT | D,D 984 | XP_006719233.1 | |
XM_017020264.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1128 | XP_016875753.1 | |
XM_017020265.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1118 | XP_016875754.1 | |
XM_017020266.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1111 | XP_016875755.1 | |
XM_017020267.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1104 | XP_016875756.1 | |
XM_017020268.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1082 | XP_016875757.1 | |
XM_017020269.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1077 | XP_016875758.1 | |
XM_017020270.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1064 | XP_016875759.1 | |
XM_017020271.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1055 | XP_016875760.1 | |
XM_017020272.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1031 | XP_016875761.1 | |
XM_017020273.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1014 | XP_016875762.1 | |
XM_017020274.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1008 | XP_016875763.1 | |
XM_017020275.1 | 2325 | Silent Mutation | GAC,GAT | D,D 1004 | XP_016875764.1 | |
XM_017020276.1 | 2325 | Silent Mutation | GAC,GAT | D,D 999 | XP_016875765.1 | |
XM_017020277.1 | 2325 | Silent Mutation | GAC,GAT | D,D 992 | XP_016875766.1 | |
XM_017020278.1 | 2325 | Silent Mutation | GAC,GAT | D,D 980 | XP_016875767.1 | |
XM_017020279.1 | 2325 | Silent Mutation | GAC,GAT | D,D 746 | XP_016875768.1 |