Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606568 | ||||||||||||||||||||
Literature Links: |
LZTFL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LZTFL1 - leucine zipper transcription factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276378.1 | 993 | UTR 3 | NP_001263307.1 | |||
NM_001276379.1 | 993 | UTR 3 | NP_001263308.1 | |||
NM_020347.3 | 993 | UTR 3 | NP_065080.1 | |||
XM_006713207.2 | 993 | UTR 3 | XP_006713270.2 | |||
XM_011533838.2 | 993 | UTR 3 | XP_011532140.1 | |||
XM_017006644.1 | 993 | UTR 3 | XP_016862133.1 | |||
XM_017006645.1 | 993 | UTR 3 | XP_016862134.1 |