Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600860 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTF3A - general transcription factor IIIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002097.2 | Intron | NP_002088.2 |
MTIF3 - mitochondrial translational initiation factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166261.1 | Intron | NP_001159733.1 | ||||
NM_001166262.1 | Intron | NP_001159734.1 | ||||
NM_001166263.1 | Intron | NP_001159735.1 | ||||
NM_152912.4 | Intron | NP_690876.3 | ||||
XM_006719771.3 | Intron | XP_006719834.1 | ||||
XM_006719772.3 | Intron | XP_006719835.1 | ||||
XM_011534957.2 | Intron | XP_011533259.1 | ||||
XM_011534958.2 | Intron | XP_011533260.1 | ||||
XM_011534959.2 | Intron | XP_011533261.1 | ||||
XM_011534960.2 | Intron | XP_011533262.1 | ||||
XM_011534961.2 | Intron | XP_011533263.1 | ||||
XM_011534962.2 | Intron | XP_011533264.1 | ||||
XM_011534963.2 | Intron | XP_011533265.1 | ||||
XM_017020417.1 | Intron | XP_016875906.1 |
Set Membership: |
HapMap |