Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602663 MIM: 602206 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRLH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PRLH - prolactin releasing hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB17 - RAB17, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022449.3 | 1683 | UTR 3 | NP_071894.1 | |||
XM_006712689.1 | 1683 | UTR 3 | XP_006712752.1 | |||
XM_017004693.1 | 1683 | Intron | XP_016860182.1 | |||
XM_017004694.1 | 1683 | Intron | XP_016860183.1 |