Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610786 | ||||||||||||||||||||||||||||||||
Literature Links: |
SRCIN1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SRCIN1 - SRC kinase signaling inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025248.2 | Intron | NP_079524.2 | ||||
XM_017025169.1 | Intron | XP_016880658.1 | ||||
XM_017025170.1 | Intron | XP_016880659.1 | ||||
XM_017025171.1 | Intron | XP_016880660.1 | ||||
XM_017025172.1 | Intron | XP_016880661.1 | ||||
XM_017025173.1 | Intron | XP_016880662.1 | ||||
XM_017025174.1 | Intron | XP_016880663.1 |