Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTCTTCAGGTTCTTCTCCTGG[A/T]AAGGCGGAGGACACACCAAACTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602941 MIM: 118890 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCAR1 - BCAR1, Cas family scaffolding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170714.1 | 3572 | UTR 3 | NP_001164185.1 | |||
NM_001170715.1 | 3572 | UTR 3 | NP_001164186.1 | |||
NM_001170716.1 | 3572 | UTR 3 | NP_001164187.1 | |||
NM_001170717.1 | 3572 | UTR 3 | NP_001164188.1 | |||
NM_001170718.1 | 3572 | UTR 3 | NP_001164189.1 | |||
NM_001170719.1 | 3572 | UTR 3 | NP_001164190.1 | |||
NM_001170720.1 | 3572 | UTR 3 | NP_001164191.1 | |||
NM_001170721.1 | 3572 | UTR 3 | NP_001164192.1 | |||
NM_014567.3 | 3572 | UTR 3 | NP_055382.2 | |||
XM_005256258.4 | 3572 | UTR 3 | XP_005256315.1 | |||
XM_005256259.4 | 3572 | UTR 3 | XP_005256316.1 | |||
XM_005256260.4 | 3572 | UTR 3 | XP_005256317.1 | |||
XM_011523475.2 | 3572 | UTR 3 | XP_011521777.1 | |||
XM_017023897.1 | 3572 | UTR 3 | XP_016879386.1 | |||
XM_017023898.1 | 3572 | UTR 3 | XP_016879387.1 | |||
XM_017023899.1 | 3572 | UTR 3 | XP_016879388.1 |
CTRB1 - chymotrypsinogen B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100506281 - uncharacterized LOC100506281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |