Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IPO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IPO4 - importin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM9SF1 - transmembrane 9 superfamily member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014842.2 | 1593 | Missense Mutation | GAC,GTC | D,V 464 | NP_001014842.1 | |
NM_001289006.1 | 1593 | Missense Mutation | GAC,GTC | D,V 377 | NP_001275935.1 | |
NM_006405.6 | 1593 | Missense Mutation | GAC,GTC | D,V 464 | NP_006396.2 |