Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSH - cathepsin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319137.1 | 1168 | UTR 3 | NP_001306066.1 | |||
NM_004390.4 | 1168 | UTR 3 | NP_004381.2 | |||
XM_005254181.2 | 1168 | UTR 3 | XP_005254238.1 | |||
XM_011521276.1 | 1168 | Intron | XP_011519578.1 | |||
XM_017021951.1 | 1168 | UTR 3 | XP_016877440.1 | |||
XM_017021952.1 | 1168 | UTR 3 | XP_016877441.1 |