Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116820 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSH PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTSH - cathepsin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319137.1 | 1205 | UTR 3 | NP_001306066.1 | |||
NM_004390.4 | 1205 | UTR 3 | NP_004381.2 | |||
XM_005254181.2 | 1205 | UTR 3 | XP_005254238.1 | |||
XM_011521276.1 | 1205 | Intron | XP_011519578.1 | |||
XM_017021951.1 | 1205 | UTR 3 | XP_016877440.1 | |||
XM_017021952.1 | 1205 | UTR 3 | XP_016877441.1 |
Set Membership: |
HapMap |