Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGATGCCGTGCTCACCCTCACG[C/T]GCCACTGTCCCTCTGAAGGCAGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCY2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCY2D - guanylate cyclase 2D, retinal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000180.3 | 937 | Missense Mutation | CGC,TGC | R,C 313 | NP_000171.1 | |
XM_011523816.1 | 937 | Missense Mutation | CGC,TGC | R,C 313 | XP_011522118.1 |