Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610976 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3H PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOBEC3H - apolipoprotein B mRNA editing enzyme catalytic subunit 3H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166002.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | NP_001159474.2 | |
NM_001166003.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | NP_001159475.2 | |
NM_001166004.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | NP_001159476.2 | |
NM_181773.4 | 200 | Missense Mutation | CGC,CTC | R,L 18 | NP_861438.3 | |
XM_011529990.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | XP_011528292.1 | |
XM_011529991.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | XP_011528293.1 | |
XM_011529992.2 | 200 | Missense Mutation | CGC,CTC | R,L 18 | XP_011528294.1 |
Set Membership: |
HapMap |