Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172860 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SERPINF1 - serpin family F member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMYD4 - SET and MYND domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052928.2 | 4396 | UTR 3 | NP_443160.2 | |||
XM_011523632.2 | 4396 | Intron | XP_011521934.1 | |||
XM_011523633.2 | 4396 | Intron | XP_011521935.1 | |||
XM_017024121.1 | 4396 | UTR 3 | XP_016879610.1 |